Learn More
Invitrogen™ HTR2B Polyclonal Antibody
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Horse, Pig and Cat.
Specifications
Specifications
| Antigen | HTR2B |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2-7.4 |
| Gene | HTR2B |
| Gene Accession No. | P41595 |
| Gene Alias | 5-HT 2B receptor; 5-HT(2B); 5-HT2B; 5-HT-2B; 5HT2B Receptor; 5-HT2b receptor; 5-HT-2F; 5-hydroxytryptamine (serotonin) receptor 2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; AJ012488; AV377389; HTR2B; NP75 protein; serotonin receptor 2B; Srl; Stomach fundus serotonin receptor |
| Gene Symbols | HTR2B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.