Learn More
HTR2B Polyclonal Antibody, Invitrogen™
Description
PA1-32494 detects 5-HT2B receptor from human samples.
Specifications
Specifications
| Antigen | HTR2B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | HTR2B |
| Gene Accession No. | P30994, P41595 |
| Gene Alias | 5-HT 2B receptor; 5-HT(2B); 5-HT2B; 5-HT-2B; 5HT2B Receptor; 5-HT2b receptor; 5-HT-2F; 5-hydroxytryptamine (serotonin) receptor 2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; AJ012488; AV377389; HTR2B; NP75 protein; serotonin receptor 2B; Srl; Stomach fundus serotonin receptor |
| Gene Symbols | HTR2B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.