Learn More
Invitrogen™ HTR2C Recombinant Rabbit Monoclonal Antibody (9H8)

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | HTR2C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 9H8 |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 |
| Gene | HTR2C |
| Gene Accession No. | P28335 |
| Gene Alias | 5ht1c; 5-HT1C; 5-HT-1C; 5HT-1C; 5-HT1c receptor; 5-HT2C; 5-HT-2C; 5HT2C Receptor; 5-HT2C receptor; 5-HT2cR; 5HTR2C; 5-HTR2C; 5-hydroxytryptamine (serotonin) receptor 2C; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; HGNC:5295; HTR1C; Htr2c; RP11-810O3.1; RP23-15B7.1; serotonin 1c receptor; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; serotonin receptor 2C; SR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.