Learn More
Invitrogen™ HTR2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 720264
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Horse, Pig, Goat, Cat and Rabbit.
Serotonin (5-hydroxytryptamine, 5-HT), a neurotransmitter, elicits a wide array of physiological effects by binding to several receptor subtypes, including the 5-HT2 family of seven-transmembrane-spanning, G-protein-coupled receptors, which activate phospholipase C and D signaling pathways. This gene encodes the 2C subtype of serotonin receptor and its mRNA is subject to multiple RNA editing events, where genomically encoded adenosine residues are converted to inosines. RNA editing is predicted to alter amino acids within the second intracellular loop of the 5-HT2C receptor and generate receptor isoforms that differ in their ability to interact with G proteins and the activation of phospholipase C and D signaling cascades, thus modulating serotonergic neurotransmission in the central nervous system. Studies in humans have reported abnormalities in patterns of 5-HT2C editing in depressed suicide victims.
Specifications
HTR2C | |
Polyclonal | |
Unconjugated | |
HTR2C | |
5ht1c; 5-HT1C; 5-HT-1C; 5HT-1C; 5-HT1c receptor; 5-HT2C; 5-HT-2C; 5HT2C Receptor; 5-HT2C receptor; 5-HT2cR; 5HTR2C; 5-HTR2C; 5-hydroxytryptamine (serotonin) receptor 2C; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; HGNC:5295; HTR1C; Htr2c; RP11-810O3.1; RP23-15B7.1; serotonin 1c receptor; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; serotonin receptor 2C; SR1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
15560, 3358 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 | |
P28335, P34968 | |
HTR2C | |
Protein corresponding to Human HTR2C (aa 236-311). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.