Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTR3A Mouse anti-Human, Clone:3B11C10, Abnova™
Mouse monoclonal antibody raised against recombinant human HTR3A.
Supplier: Abnova Corporation MAB17568
Description
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit A of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]Specifications
HTR3A | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry (1:100-1:500) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. |
|
HTR3A | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3B11C10 | |
5-hydroxytryptamine (serotonin) receptor 3A | |
In PBS (0.05% sodium azide) | |
5-HT-3, 5-HT3A, 5-HT3R, 5HT3R, HTR3 | |
Recombinant protein corresponding to amino acid 24-157 of human HTR3A from E. coli. | |
3359 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction