Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01676642
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-642 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-642 Supplier Invitrogen™ Supplier No. PA141033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human brain tissue, human brain protein. Immunogen sequence: RATQAHSTTQ PALLRL & LSSIRHSLEK RDEMRE.

The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit A of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Htr3a
Gene Accession No. P23979, P35563, P46098
Gene Alias 5HT3; 5-HT3; 5-HT-3; 5-HT3 receptor; 5HT3 serotonin receptor; 5-HT3A; 5-HT3-A; 5HT3R; 5-HT3R; 5-hydroxytryptamine (serotonin) receptor 3A; 5-hydroxytryptamine (serotonin) receptor 3A, ionotropic; 5-hydroxytryptamine receptor 3; 5-hydroxytryptamine receptor 3A; Htr3; Htr3a; serotonin receptor; Serotonin receptor 3A; serotonin-gated ion channel receptor; truncated receptor
Gene Symbols Htr3a
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 27-42 and 427-442 of rat 5-HT3 Receptor.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15561, 3359, 79246
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.