Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR3B Monoclonal Antibody (4G6H7)
GREENER_CHOICE

Catalog No. PIMA531773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531773 Supplier Invitrogen™ Supplier No. MA531773
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31773 has been tested in indirect ELISA.

The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit B of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR3B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G6H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene HTR3B
Gene Accession No. O95264
Gene Alias 5-HT3 receptor subunit B; 5-HT3B; 5-HT-3B; 5-HT3-B; 5-HT3B receptor; 5-hydroxytryptamine (serotonin) receptor 3B; 5-hydroxytryptamine (serotonin) receptor 3B, ionotropic; 5-hydroxytryptamine 3 receptor B subunit; 5-hydroxytryptamine receptor 3B; HTR3B; Serotonin receptor 3B; serotonin-gated ion channel subunit
Gene Symbols HTR3B
Host Species Mouse
Immunogen Purified recombinant fragment of human HTR3B (AA: extra 23-150) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.