Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR3B Mouse anti-Human, Clone:2H9H5, Abnova™

Catalog No. 89334036 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-036 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-036 Supplier Abnova Corporation Supplier No. MAB17570
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human HTR3B.

The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit B of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. [provided by RefSeq]

Specifications

Antigen HTR3B
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2H9H5
Conjugate Unconjugated
Description 5-hydroxytryptamine (serotonin) receptor 3B
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry (1:100-1:500)
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene HTR3B
Gene Alias 5-HT3B
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 23-150 of human HTR3B from E. coli.
Quantity 100 μg
Gene ID (Entrez) 9177
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.