Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HTR3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541033
Description
Peptide sequence: PLWACILVAA GILATDTHHP QDSALYHLSK QLLQKYHKEV RPVYNWTKAT Sequence homology: Cow: 79%; Dog: 77%; Guinea Pig: 79%; Human: 100%; Mouse: 77%; Pig: 86%; Rabbit: 86%; Rat: 79%.
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit B of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude.
Specifications
| HTR3B | |
| Polyclonal | |
| Unconjugated | |
| HTR3B | |
| 5-HT3 receptor subunit B; 5-HT3B; 5-HT-3B; 5-HT3-B; 5-HT3B receptor; 5-hydroxytryptamine (serotonin) receptor 3B; 5-hydroxytryptamine (serotonin) receptor 3B, ionotropic; 5-hydroxytryptamine 3 receptor B subunit; 5-hydroxytryptamine receptor 3B; HTR3B; Serotonin receptor 3B; serotonin-gated ion channel subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57014, 9177 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95264, Q9JHJ5 | |
| HTR3B | |
| Synthetic peptide directed towards the N-terminal of human HTR3B (aa 8-57). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction