Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HTR3C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525865
Description
HTR3C Polyclonal Antibody for Western Blot
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit C of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3.
Specifications
| HTR3C | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8WXA8 | |
| HTR3C | |
| KLH conjugated synthetic peptide between 128-157 amino acids from the central region of human HTR3C. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| HTR3C | |
| 5-HT3C; 5-HT3-C; 5-hydroxytryptamine (serotonin) receptor 3, family member C; 5-hydroxytryptamine (serotonin) receptor 3C, ionotropic; 5-hydroxytryptamine receptor 3 subunit C; 5-hydroxytryptamine receptor 3C; HTR3C; Serotonin receptor 3C | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 170572 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction