Learn More
Invitrogen™ HTR3D Polyclonal Antibody

Description
HTR3D Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | HTR3D |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | HTR3D |
| Gene Accession No. | Q70Z44 |
| Gene Alias | 5HT3D; 5-HT3-D; 5-hydroxytryptamine (serotonin) receptor 3 family member D; 5-hydroxytryptamine (serotonin) receptor 3D, ionotropic; 5-hydroxytryptamine receptor 3 subunit D; 5-hydroxytryptamine receptor 3D; HTR3D; serotonin 5-HT-3D receptor; serotonin receptor 3D |
| Gene Symbols | HTR3D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.