Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR3E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562036 Supplier Invitrogen™ Supplier No. PA562036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAFILSRATP RPALGPLSYR EHRVALLHLT HSMSTTGRGV TFTINCSGF Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 27%.

The product of this gene belongs to the ligand-gated ion channel receptor superfamily. HTR3E encodes a subunit E of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3. An alternative splice variant has been described but its full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR3E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HTR3E
Gene Accession No. A5X5Y0
Gene Alias 5-HT3c1; 5-HT3E; 5-HT3-E; 5-hydroxytryptamine (serotonin) receptor 3, family member E; 5-hydroxytryptamine (serotonin) receptor 3E, ionotropic; 5-hydroxytryptamine receptor 3 subunit E; 5-hydroxytryptamine receptor 3E; HTR3E; Serotonin receptor 3E
Gene Symbols HTR3E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HTR3E. Recombinant protein control fragment (Product #RP-99958).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285242
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.