Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HTR3E Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568698
Description
This target displays homology in the following species: Dog: 100%; Horse: 83%; Human: 100%; Rabbit: 83%.
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. HTR3E encodes a subunit E of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3. An alternative splice variant has been described but its full length sequence has not been determined.
Specifications
| HTR3E | |
| Polyclonal | |
| Unconjugated | |
| HTR3E | |
| 5-HT3c1; 5-HT3E; 5-HT3-E; 5-hydroxytryptamine (serotonin) receptor 3, family member E; 5-hydroxytryptamine (serotonin) receptor 3E, ionotropic; 5-hydroxytryptamine receptor 3 subunit E; 5-hydroxytryptamine receptor 3E; HTR3E; Serotonin receptor 3E | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 285242 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A5X5Y0 | |
| HTR3E | |
| synthetic peptide directed towards the middle region of human HTR3E. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction