Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532867
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532867 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532867 Supplier Invitrogen™ Supplier No. PA532867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Elephant, Panda, Horse, Guinea pig (100%); Opossum (94%); Turkey, Chicken (89%); Lizard, Zebrafish, Stickleback (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen. This antibody is not recommended for use in WB.

The neurotransmitter serotonin (5-hydroxytryptamine, 5-HT) has been implicated in a wide range of psychiatric conditions and also has vasoconstrictive and vasodilatory effects. The gene described in this record is a member of 5-hydroxytryptamine (serotonin) receptor family and encodes a multi-pass membrane protein that functions as a receptor for 5-hydroxytryptamine and couples to G-proteins. This protein has been shown to function in part through the regulation of intracellular Ca2+ mobilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR5A
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene HTR5A
Gene Accession No. P30966, P35364, P47898
Gene Alias 5-HT-5; 5HT5A; 5-HT5A; 5-HT-5A; 5-HT5a receptor; 5-hydroxytryptamine (serotonin) receptor 5A; 5-hydroxytryptamine (serotonin) receptor 5A, G protein-coupled; 5-hydroxytryptamine receptor 5A; 5-hydroxytryptamine5a; Htr5; Htr5a; MR22; REC17; Serotonin receptor 5A
Gene Symbols HTR5A
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 1st extracellular domain of human 5HT5A Receptor.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100063709, 100379278, 15563, 25689, 3361, 482814, 541200
Target Species Human, Mouse, Rat, Canine, Monkey, Bovine, Horse, Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.