Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HTR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533854
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset, Rat, Hamster, Panda, Dog, Horse (100%).
This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.
Specifications
| HTR6 | |
| Polyclonal | |
| Unconjugated | |
| HTR6 | |
| 5-HT6; 5-HT-6; 5-HT6 receptor; 5-HT6R; 5-hydroxytryptamine (serotonin) receptor 6; 5-hydroxytryptamine (serotonin) receptor 6, G protein-coupled; 5-hydroxytryptamine receptor 6; Htr6; Serotonin receptor 6; ST-B17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100058656, 100752934, 3362, 487402, 64354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 0.1% sodium azide | |
| P31388, P50406 | |
| HTR6 | |
| Synthetic 17 amino acid peptide from C-terminus of human HTR6 / 5-HT6. | |
| 50 μg | |
| Primary | |
| Human, Rat, Canine, Monkey, Hamster, Horse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction