Learn More
Invitrogen™ HTR7 Polyclonal Antibody

Description
PA1-24575 detects 5-HT7 Receptor in human samples.
Specifications
Specifications
| Antigen | HTR7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | HTR7 |
| Gene Accession No. | P32304, P34969 |
| Gene Alias | 5-HT receptor 7a; 5-HT receptor 7b; 5Ht7; 5-HT7; 5-HT-7; 5HT7 Receptor; 5-HT7 receptor; 5-HT-X; 5-hydroxytryptamine (serotonin) receptor 7; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine receptor 7; GPRFO; high affinity serotonin receptor (5HT7); HT7; Htr7; OTTHUMP00000020077; OTTHUMP00000020078; OTTHUMP00000020079; Serotonin receptor 7 |
| Gene Symbols | HTR7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.