Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675704
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-704 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-704 Supplier Invitrogen™ Supplier No. PA141122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with porcine samples based on sequence homology. The immunogen sequence is 93% conserved with canine 5-HT7Ra/b, and 81% conserved with porcine 5-HT7R.

5-HT7 Receptor is a Serotonin Receptor with several functions, including regulation of the circadian rhythm and mediation of smooth-muscle relaxation in the vasculature and GI tract. This receptor has been implicated in depression, autism, and other neuropsychiatric disorders. Three protein isoforms have been documented: isoform a: 445aa; isoform b: 432aa; isoform d: 479aa. 5-HT7 Receptor has been reported in aorta, brain, heart, placenta, small intestine, spleen, artery, and vessel. An EST has been isolated from a testis library.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene HTR7
Gene Accession No. P32304, P32305, P34969
Gene Alias 5-HT receptor 7a; 5-HT receptor 7b; 5Ht7; 5-HT7; 5-HT-7; 5HT7 Receptor; 5-HT7 receptor; 5-HT-X; 5-hydroxytryptamine (serotonin) receptor 7; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine receptor 7; GPRFO; high affinity serotonin receptor (5HT7); HT7; Htr7; OTTHUMP00000020077; OTTHUMP00000020078; OTTHUMP00000020079; Serotonin receptor 7
Gene Symbols HTR7
Host Species Rabbit
Immunogen Mixture of synthetic peptides corresponding to amino acids 13-28 of the rat 5-HT7R.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15566, 3363, 477762, 65032
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.