Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTRA2/Omi Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP29940950UL
Description
HTRA2/Omi Polyclonal antibody specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
HTRA2/Omi | |
Polyclonal | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 uL/mg of lysate | |
High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
Produced in rabbits immunized with purified, recombinant Human HTRA2/Omi (Accession#: O43464-1; Ala134-Glu458) | |
50 μg | |
Apoptosis, Cancer, Mitochondrial Mediated Pathway | |
27429 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction