Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTRA2/Omi Antibody (ST54-05), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267543
Description
HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HTRA2/Omi | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
Rabbit | |
Protein A purified | |
Apoptosis, Cancer, Mitochondrial Mediated Pathway | |
27429 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
ST54-05 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
HTRA2 | |
Recombinant protein within Human HTRA2/Omi aa 800-188/ 458. (SwissProt: O43464 Human; SwissProt: Q9JIY5 Mouse; Unigene: 107325 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction