Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HTRA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590098
Description
Immunogen sequence: PSDRLREFLH RGEKKNSSSG ISGSQRRYIG VMMLTLSPSI LAELQLREPS FPDVQHGVLI HKVILGSPAH RAGLRPGDVI LAIGEQMVQN AEDVYEAVRT QSQLAVQIRR GRETLTLYVT PEVTE.
The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity (1-3). Recently, a serine protease has been isolated, which is released from the mitochondria upon induction of apoptosis by apoptotic stimuli. This protein called Omi/HtrA2 is a mitochondrial protein that binds to IAP (4). Like Smac/DIABLO, the mature Omi protein contains a conserved IAP-binding motif (AVPS) at its N terminus. The deregulated expression of Omi in the cytoplasm of mammalian cells induces apoptosis indicating that Omi could participate in the mitochondrial apoptotic pathway.
Specifications
HTRA2 | |
Polyclonal | |
Unconjugated | |
HTRA2 | |
AI481710; High temperature requirement protein A2; HtrA serine peptidase 2; HtrA2; HtrA-like serine protease; mnd2; motor neuron degeneration 2; OMI; Omi stress-regulated endoprotease; PARK13; protease, serine, 25; Prss25; Serine protease 25; serine protease HTRA2, mitochondrial; serine protease OMI; serine proteinase OMI | |
Rabbit | |
Affinity chromatography | |
RUO | |
27429, 297376, 64704 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43464, Q9JIY5 | |
HTRA2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 334-458 of human HTRA2 (NP_0373791). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction