Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HtrA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | HtrA3 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HtrA3 Polyclonal specifically detects HtrA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HtrA3 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
94031 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.108, High-temperature requirement factor A3, HtrA serine peptidase 3, Pregnancy-related serine protease, probable serine protease HTRA3, Prsp, serine protease HTRA3, Tasp | |
Recombinant fusion protein containing a sequence corresponding to amino acids 334-453 of human HTRA3 (NP_444272.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title