Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
htrA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169684
Description
htrA4 Polyclonal specifically detects htrA4 in Human samples. It is validated for Western Blot.Specifications
htrA4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.108, FLJ90724, HtrA serine peptidase 4, probable serine protease HTRA4 | |
Rabbit | |
51 kDa | |
100 μL | |
Primary | |
Guinea pig: 86%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
P83105 | |
HTRA4 | |
Synthetic peptides corresponding to HTRA4(HtrA serine peptidase 4) The peptide sequence was selected from the middle region of HTRA4 (NP_710159). Peptide sequence LKMHYPDFPDVSSGVYVCKVVEGTAAQSSGLRDHDVIVNINGKPITTTTD. | |
Affinity purified | |
RUO | |
203100 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction