Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTT Mouse anti-Human, Clone: 3-19, Abnova™

Catalog No. 89035499 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-499 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-499 Supplier Abnova Corporation Supplier No. MAB14101

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against synthetic peptide of human HTT.

  • High specificity
  • High reactivity

Specifications

Antigen HTT
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3-19
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human HTT.
Dilution ELISA (1:100) Immunofluorescence (1:500) Immunohistochemistry (1:500) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P42858
Gene Alias HD/IT15
Gene Symbols HTT
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 505-513 of human HTT.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3064
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.