Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Htz1 / Histone H2A.Z Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$159.90 - $633.45

Specifications

Antigen Htz1 / Histone H2A.Z
Concentration Lot-Specific
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2758
SDP
Encompass
View Documents
Active Motif
39648
10 μL
Each for $159.90
Only null left
Add to Cart
 
50-199-2757
SDP
Encompass
View Documents
Active Motif
39647
100 μL
Each for $633.45
Only null left
Add to Cart
 
Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.
Specifications

Specifications

Htz1 / Histone H2A.Z
ChIP Assay, Western Blot
Unconjugated
Rabbit
Yeast
11ac; BOS_6451; C530002L11Rik; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member V; H2A histone family, member Z; H2A.F/Z; H2A.Z; H2A.Z1; H2A.Z-1; H2a.z-2; H2A.ZK5; H2A.ZK5ac; H2A.Zl; h2a.zl1; h2a.zl2; H2A/z; H2afv; H2afz; h2afz.L; H2AFZac; H2av; H2AZ; H2AZ histone; H2AZK11ac; H2AZK7ac; histone H2A.F/Z; Histone H2A.V; Histone H2A.Z; histone H2A.Zl2; Histone H2A.Z-like; XELAEV_18005732mg; XH2AZ
IgG
Antigen affinity chromatography
Antibody
Lot-Specific
Polyclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
This Htz1 antibody was raised against full-length Htz1 protein from S. cerevisiae.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.