Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AKT (Phospho) [pS473] ELISA Kit

Catalog No. kho0111
Encompass_Preferred
Click to view available options
Quantity:
96 Tests

ELISA

The Human AKT (Phospho) [pS473] ELISA Kit is designed to detect and quantify the level of AKT (Phospho) [pS473] in fresh or frozen human cell lysates. The assay recognizes both natural and recombinant AKT (Phospho) [pS473]. Principle of the method A monoclonal capture antibody specific for AKT (Phospho) [pS473] has been coated onto the wells of the 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a horseradish peroxidase labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

AKT1 (PKB alpha) is a serine/threonine kinase that regulates cell survival. The activated enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P31749
Assay Range 1.6 to 100 U/mL
Assay Sensitivity <0.8 U/mL
Concentration 1.6 U/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Substrate Type Serine/Threonine Kinases
Target Species Rat, Mouse, Human
For Use With (Application) Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Drug Discovery and Development, Immunoassays, Kinase Antibodies and Immunoassays, Kinase Biology, Ready-To-Use Immunoassay, Target and Lead Identification and Validation
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 207
Gene Symbol AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA,HIHGHH,PKBB,PKBBETA,PRKBB,RAC-BETA,MPPH,MPPH2,PKB-GAMMA,PKBG,PRKBG,RAC-Pk.-gamma,RAC-gamma,STK-2
Incubation Time 4 hr.
Interassay CV 8.3%
Intraassay CV 6.9
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene Akt
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL
Storage Requirements 2°C to 8°C
Target AKT1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.