Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD183 Mouse anti-Human, Rhesus, Cynomolgus, Baboon, BB700, Clone: 1C6/CXCR3 (also known as 1C6, LS177-1C6), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566533
Description
CD183 Monoclonal antibody specifically detects CD183 in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Flow Cytometry
Specifications
CD183 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
CXCR3; C-X-C chemokine receptor type 3; GPR9; IP10R; MigR; CKRL2; CMKAR3 | |
Human CXCR3 Peptide | |
RUO | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
IgG1 κ |
Flow Cytometry | |
1C6/CXCR3 (also known as 1C6, LS177-1C6) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB700 under optimum conditions, and unconjugated antibody and free BD Horizon BB700 were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction