Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD33 Mouse anti-Human, BUV496, Clone: WM53 (also known as WM-53), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 741148
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow Cytometry
Specifications
CD33 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
Siglec-3; SIGLEC3; Sialic acid-binding Ig-like lectin 3; p67; gp67; My9 | |
Acute Myeloid Leukemia Blasts | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
WM53 (also known as WM-53) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction