Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Mouse anti-Human , BV786, Clone: OKT4, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566807
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry
Specifications
CD4 | |
Monoclonal | |
Brilliant Violet 786 | |
CD4 antigen (p55); T-cell surface antigen T4/Leu-3 | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
OKT4 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction