Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10 Rat anti-Human, Virus, BB700, Clone: JES3-19F1, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566567
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human,Virus samples. It is validated for Flow Cytometry.
Specifications
IL-10 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Human, Virus | |
IgG2a κ |
Flow Cytometry | |
JES3-19F1 | |
Rat | |
RUO | |
IL-10 | |
Refrigerate (2 to 8°C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction