Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HuA/B/C/D Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
SDP

Catalog No. 501728433 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8433 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8433 Supplier Proteintech Group Inc Supplier No. 130321AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Embryonic lethal, abnormal vision, Drosophila-like 4 (ELAVL4) is a human homologue of the Drosophila gene ELAV, which is implicated in neuronal differentiation and maintece. The ELAVL proteins are mRNA binding proteins and all members of the family contain three RNA-recognition motifs. ELAVL-proteins also interact with inflammation-associated factors. The protein may be implicated in neurodegeneration through these complex mechanisms.

Specifications

Antigen HuA/B/C/D
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ELAVL1
Gene Accession No. B5DF91, O09032, P26378, P70372, Q12926, Q14576, Q15717, Q60899, Q60900, Q61701, Q8CH84
Gene Alias ELAV like protein 4, ELAVL4, Hu antigen D, Hu C/D, HUD, PNEM
Gene Symbols Elavl1, elavl1a, Elavl2, Elavl3, Elavl4
Host Species Rabbit
Immunogen HuA/B/C/D Fusion Protein Ag3678
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15568, 15569, 15571, 15572, 1993, 1994, 1995, 1996, 282824, 286973, 30732, 30736, 30737, 363854, 431719, 432358
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.