Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HuC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501735949 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5949 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5949 Supplier Proteintech Group Inc Supplier No. 550471AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody has no cross reaction to other ELAVL members.

HUC, belongs to the RRM elav family. It binds to AU-rich sequences (AREs) of target mRNAs, including VEGF mRNA. ELAVL3 may also bind poly-A tracts via RRM 3. It is involved in neuronal differentiation and maintece.

Specifications

Antigen HuC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ELAVL3
Gene Accession No. Q14576, Q60900
Gene Alias ELAV like protein 3, ELAVL3, Hu antigen C, HUC, HUCL, PLE21
Gene Symbols ELAVL3
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15571, 1995, 282824
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.