Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 11 beta-HSD2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8630SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8630SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8630SP Supplier R&D Systems Supplier No. MAB8630SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSD11B2 Monoclonal specifically detects HSD11B2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HSD11B2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 921215
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P80365
Gene Alias 11 betaHSD2, AME, corticosteroid 11-beta-dehydrogenase isozyme 2,11-beta-HSD2, EC 1.1.1, EC 1.1.1.-, HSD11B2, HSD11KAME1, HSD2,11-DH2, hydroxysteroid (11-beta) dehydrogenase 2, NAD-dependent 11-beta-hydroxysteroid dehydrogenase, SDR9C3, short chain dehydrogenase/reductase family 9C member 3,11-beta-hydroxysteroid dehydrogenase type 2
Gene Symbols HSD11B2
Host Species Mouse
Immunogen E. coli-derived recombinant human 11 beta-HSD2 Met105-Arg405 Accession # P80365
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3291
Test Specificity Detects human 11 beta-HSD2 in direct ELISA and Western Blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.