Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 14-3-3 zeta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2669SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2669SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2669SP Supplier R&D Systems Supplier No. MAB2669SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

14-3-3 zeta Monoclonal specifically detects 14-3-3 zeta in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen 14-3-3 zeta
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 818515
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P63104
Gene Alias 1433 zeta, KCIP-1, KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, YWHAZ, zeta polypeptide
Gene Symbols YWHAZ
Host Species Mouse
Immunogen E. coli-derived recombinant human 14-3-3 zeta Asp2-Asn245 Accession # P63104
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7534
Test Specificity Detects human 14-3-3 zeta in direct ELISAs and Western blots. Detects Mouse and Rat 14-3-3 zeta in Western Blots. In direct ELISAs, no cross-reactivity with recombinant human 14-3-3 beta, theta, eta, gamma, sigma, or epsilon is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.