Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 17 beta-HSD1/HSD17B1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7178SP
This item is not returnable.
View return policy
Description
17 beta-HSD1/HSD17B1 Polyclonal specifically detects 17 beta-HSD1/HSD17B1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
17 beta-HSD1/HSD17B1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
17 betaHSD1, 17-beta-HSD 1, 17-beta-hydroxysteroid dehydrogenase type 1, 20 alpha-hydroxysteroid dehydrogenase, 20-alpha-HSD, E17KSR, E2DH, EC 1.1.1.62, EDH17B1, EDH17B2, EDH17B2EDHB17, EDHB17, estradiol 17-beta-dehydrogenase 1, estradiol 17-beta-dehydrogenase-1, HSD17, HSD17B1, hydroxysteroid (17-beta) dehydrogenase 1, hydroxysteroid (17-beta) dehydrogenase 1 isoform, MGC138140, Placental 17-beta-hydroxysteroid dehydrogenase, SDR28C1, short chain dehydrogenase/reductase family 28CE, member 1 | |
Sheep | |
Affinity Purified | |
RUO | |
3292 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.2 μg/mL, Simple Western 2.5 μg/mL, Immunohistochemistry 5-15 μg/mL | |
P14061 | |
HSD17B1 | |
E. coli-derived recombinant human 17 beta-HSD1/HSD17B1 Ala2-Gln328 Accession # P14061 | |
25 μg | |
Primary | |
Detects human 17 beta-HSD1/HSD17B1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction