Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 17 beta-HSD1/HSD17B1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7178
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7178 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7178 Supplier R&D Systems Supplier No. AF7178
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

17 beta-HSD1/HSD17B1 Polyclonal specifically detects 17 beta-HSD1/HSD17B1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen 17 beta-HSD1/HSD17B1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 μg/mL, Simple Western 2.5 μg/mL, Immunohistochemistry 5-15 μg/mL
Gene Accession No. P14061
Gene Alias 17 betaHSD1, 17-beta-HSD 1, 17-beta-hydroxysteroid dehydrogenase type 1, 20 alpha-hydroxysteroid dehydrogenase, 20-alpha-HSD, E17KSR, E2DH, EC 1.1.1.62, EDH17B1, EDH17B2, EDH17B2EDHB17, EDHB17, estradiol 17-beta-dehydrogenase 1, estradiol 17-beta-dehydrogenase-1, HSD17, HSD17B1, hydroxysteroid (17-beta) dehydrogenase 1, hydroxysteroid (17-beta) dehydrogenase 1 isoform, MGC138140, Placental 17-beta-hydroxysteroid dehydrogenase, SDR28C1, short chain dehydrogenase/reductase family 28CE, member 1
Gene Symbols HSD17B1
Host Species Sheep
Immunogen E. coli-derived recombinant human 17 beta-HSD1/HSD17B1 Ala2-Gln328 Accession # P14061
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3292
Test Specificity Detects human 17 beta-HSD1/HSD17B1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.