Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 17 beta-HSD14/HSD17B14 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7515SP
This item is not returnable.
View return policy
Description
17 beta-HSD14/HSD17B14 Monoclonal specifically detects 17 beta-HSD14/HSD17B14 in Human samples. It is validated for Immunohistochemistry.Specifications
17 beta-HSD14/HSD17B14 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q9BPX1 | |
HSD17B14 | |
E. coli-derived recombinant human 17 beta-HSD14/HSD17B14 Ala2-Ser270 Accession # Q9BPX1 | |
25 μg | |
Primary | |
Detects human 17 beta-HSD14/HSD17B14 in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
736043 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
17 betaHSD14, dehydrogenase/reductase (SDR family) member 10, Dehydrogenase/reductase SDR family member 10, DHRS10, DHRS1017-beta-hydroxysteroid dehydrogenase 14, EC 1.1.1.-, HSD17B14, hydroxysteroid (17-beta) dehydrogenase 14,17-beta-hydroxysteroid dehydrogenase DHRS10, retinal short-chain dehydrogenase/reductase 3, Retinal short-chain dehydrogenase/reductase retSDR3, retSDR3, RETSDR3,17-beta-HSD 14, SDR3, SDR47C1, short chain dehydrogenase/reductase family 47C, member 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
51171 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction