Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 17 beta-HSD14/HSD17B14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7515
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7515 Supplier R&D Systems Supplier No. MAB7515
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

17 beta-HSD14/HSD17B14 Monoclonal specifically detects 17 beta-HSD14/HSD17B14 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen 17 beta-HSD14/HSD17B14
Applications Immunohistochemistry
Classification Monoclonal
Clone 736043
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BPX1
Gene Alias 17 betaHSD14, dehydrogenase/reductase (SDR family) member 10, Dehydrogenase/reductase SDR family member 10, DHRS10, DHRS1017-beta-hydroxysteroid dehydrogenase 14, EC 1.1.1.-, HSD17B14, hydroxysteroid (17-beta) dehydrogenase 14,17-beta-hydroxysteroid dehydrogenase DHRS10, retinal short-chain dehydrogenase/reductase 3, Retinal short-chain dehydrogenase/reductase retSDR3, retSDR3, RETSDR3,17-beta-HSD 14, SDR3, SDR47C1, short chain dehydrogenase/reductase family 47C, member 1
Gene Symbols HSD17B14
Host Species Mouse
Immunogen E. coli-derived recombinant human 17 beta-HSD14/HSD17B14 Ala2-Ser270 Accession # Q9BPX1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51171
Test Specificity Detects human 17 beta-HSD14/HSD17B14 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.