Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 20S Immunoproteasome Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | 20S Immunoproteasome |
---|---|
Clone | 894505 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB81941SP
![]() |
R&D Systems
MAB81941SP |
25 μg |
Each for $200.50
|
|
|||||
MAB81941100
![]() |
R&D Systems
MAB81941100 |
100 μg |
Each for $491.00
|
|
|||||
Description
20S Immunoproteasome Monoclonal specifically detects 20S Immunoproteasome in Human samples. It is validated for Immunohistochemistry.Specifications
20S Immunoproteasome | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
Human 20S Immunoproteasome purified from peripheral blood mononuclear cells | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Human 20S Immunoproteasome in direct ELISAs. |
894505 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title