Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 20S Immunoproteasome Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8194100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8194100 100 μg
MAB8194SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8194100 Supplier R&D Systems Supplier No. MAB8194100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

20S Immunoproteasome Monoclonal specifically detects 20S Immunoproteasome in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen 20S Immunoproteasome
Applications Immunocytochemistry
Classification Monoclonal
Clone 894510
Conjugate Unconjugated
Dilution Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Host Species Mouse
Immunogen Human 20S Immunoproteasome purified from peripheral blood mononuclear cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human 20S Immunoproteasome in direct ELISA.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.