Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 2B4/CD244/SLAMF4 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10393N10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10393N10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10393N10 Supplier R&D Systems Supplier No. FAB10393N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Flow Cytometry
Classification Monoclonal
Clone 999602
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. NP_057466
Gene Alias CD244, CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAIL, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4, SLAMF4NK cell activation inducing ligand NAIL
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human 2B4/CD244/SLAMF4, Gly19-His222, Accession # NP_057466
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51744
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.