Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 2B4/CD244/SLAMF4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1039SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1039SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1039SP Supplier R&D Systems Supplier No. MAB1039SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 146510
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_057466
Gene Alias CD244, CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAIL, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4, SLAMF4NK cell activation inducing ligand NAIL
Gene Symbols CD244
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human 2B4/CD244/SLAMF4 Cys22-Arg221 Accession # NP_057466
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51744
Test Specificity Detects human 2B4/CD244/SLAMF4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse 2B4/CD244/SLAMF4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.