Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 4-1BB Ligand/TNFSF9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB22951100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB22951100 100 μg
MAB22951SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB22951100 Supplier R&D Systems Supplier No. MAB22951100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen TIM-3
Applications Immunohistochemistry
Classification Monoclonal
Clone 2357C
Conjugate Unconjugated
Dilution Immunohistochemistry 3-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P41273.1
Gene Alias 4-1BB ligand, 41BB Ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Host Species Rabbit
Immunogen E. coli-derived recombinant human 4-1BB Ligand/TNFSF9, Arg71-Glu254, Accession # P41273.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 84868
Test Specificity Detects human 4-1BB Ligand/TNFSF9 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.