Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human 4-1BB Receptor Recombinant Protein, PeproTech®

Catalog No. p-7245981
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human soluble 4-1BB Receptor is a 167 amino acid polypeptide (17.7 kDa), which contains the cysteine-rich TNFR-like extracellular domain of 4-1BB Receptor. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07011
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3604
Molecular Weight (g/mol) 17.7 kDa
Name Human 4-1 BB Receptor
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 4 1 BB; 41 BB; 4-1 BB; 4-1 BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1 BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1 BB; s4 1 BB; s4 1 BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1 BB; soluble 41 BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1 BB; T-cell antigen 4-1 BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9
Common Name 4-1 BB (CD137)
Gene Symbol TNFRSF9
Biological Activity Determined by its inhibitory effect of the 4-1 BBL mediated stimulation of IL-8 production by human PBMC. About 90% of inhibition was seen using a concentration of 1 ug for both 4-1 BBL and 4-1 BBR.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MERTRSLQDP CSNCPAGTFC DNNRNQICSP CPPNSFSSAG GQRTCDICRQ CKGVFRTRKE CSSTSNAECD CTPGFHCLGA GCSMCEQDCK QGQELTKKGC KNCCFGTFND QKRGICRPWT NCSLDGKSVL VNGTKERDVV CGPSPADLSP GASSVTPPAP AREPGHS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.