Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human 4-1BB/TNFRSF9 DuoSet ELISA
SDP

Catalog No. DY838 Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
15 x 96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DY838 15 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY838 Supplier R&D Systems™ Supplier No. DY838
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human 4-1BB/TNFRSF9 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human 4-1BB/TNFRSF9 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human 4-1BB/TNFRSF9/CD137.

Specifications

Quantity 1 Kit (for 15 Plates)
Species Human
Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 3604
Synonym 41 BB, 4-1 BB, 4-1 BB ligand receptor, CD137, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILA, ILAhomolog of mouse 4-1 BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1 BB, T cell antigen ILA, T-cell antigen 4-1 BB homolog, T-cell antigen ILA, TNFRSF9, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Packaging Quantity 15 x 96-well Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.