Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 5-HT1B Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $491.00

Specifications

Antigen 5-HT1B
Clone 499325
Concentration LYOPH
Dilution Immunohistochemistry 8-25 ug/mL
Applications Immunohistochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5858SP
GREENER_CHOICE
View Documents
R&D Systems
MAB5858SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB5858
GREENER_CHOICE
View Documents
R&D Systems
MAB5858
100 μg
Each for $491.00
Only null left
Add to cart
 
Description

Description

5-HT1B Monoclonal specifically detects 5-HT1B in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

5-HT1B
LYOPH
Immunohistochemistry
Unconjugated
Mouse
Human
P28222
3351
NS0 mouse myeloma cell line transfected with human 5HT1B Met1-Ser390 Accession # P28222
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human 5HT1B.
499325
Immunohistochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
5HT1B, 5-HT1B5-HT-1B, 5HT1DB, 5-HT1DB, 5-hydroxytryptamine (serotonin) receptor 1B, HTR1B, HTR1D2, HTR1D2,5-hydroxytryptamine receptor 1B, HTR1DB, S12, S125-HT-1D-beta, Serotonin 1D beta receptor, Serotonin receptor 1B
HTR1B
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.