Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 5-HT2A Alexa Fluor™ 488-conjugated Antibody, Novus Biologicals™
GREENER_CHOICE

Catalog No. FAB2501G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2501G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2501G100 Supplier R&D Systems Supplier No. FAB112501G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

5-HT2A Monoclonal antibody specifically detects 5-HT2A in Human samples. It is validated for Flow Cytometry

Specifications

Antigen 5-HT2A
Applications Flow Cytometry
Classification Monoclonal
Clone 1055220
Conjugate Alexa Fluor 488
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P28223
Gene Alias 5-HT-2, 5-HT2 receptor, 5HT2A, 5-HT-2A, 5-hydroxytryptamine (serotonin) receptor 2A, 5-hydroxytryptamine receptor 2A, HTR2,5-HT2A, HTR2A, serotonin 5-HT-2A receptor, Serotonin receptor 2A
Host Species Mouse
Immunogen Synthetic peptide corresponding to the amino acids 1-75 in the N-term extracellular domain of the Serotonin Receptor 5-HT2A (HTR2A)., Accession # P28223
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3356
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.