Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 5-HT2A Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB112501SP
This item is not returnable.
View return policy
Description
5-HT2A Monoclonal antibody specifically detects 5-HT2A in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
5-HT2A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
5-HT-2, 5-HT2 receptor, 5HT2A, 5-HT-2A, 5-hydroxytryptamine (serotonin) receptor 2A, 5-hydroxytryptamine receptor 2A, HTR2,5-HT2A, HTR2A, serotonin 5-HT-2A receptor, Serotonin receptor 2A | |
Synthetic peptide corresponding to the amino acids 1-75 in the N-term extracellular domain of the Serotonin Receptor 5-HT2A (HTR2A)., Accession # P28223 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
1055220 | |
Flow Cytometry 0.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
P28223 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3356 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction