Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 5-HT2B APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000079
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000079 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000079 Supplier R&D Systems Supplier No. FAB11297A100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

5-HT2B Monoclonal antibody specifically detects 5-HT2B in Human samples. It is validated for Flow Cytometry

Specifications

Antigen 5-HT2B
Applications Flow Cytometry
Classification Monoclonal
Clone 2880B
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P41595
Gene Alias 5-HT(2B), 5HT2B, 5-HT-2B, 5-HT2B5-HT 2B receptor, 5-hydroxytryptamine (serotonin) receptor 2B, 5-hydroxytryptamine 2B receptor, 5-hydroxytryptamine receptor 2B, HTR2B, Serotonin receptor 2B
Host Species Rabbit
Immunogen Synthetic peptide containing human 5- HTR2B, Accession # P41595
Purification Method Protein A or G purified from ascites
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3357
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.