Learn More
Human 5-HT2C Alexa Fluor 594-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | 5-HT2C |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 496214 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Alias | 5-HT1C, 5-HT-1C, 5HT2C, 5-HT-2C, 5-HT2C5-hydroxytryptamine receptor 1C, 5-HTR2C, 5-hydroxytryptamine (serotonin) receptor 2C, 5-hydroxytryptamine receptor 2C, HTR1C, HTR2C, serotonin 5-HT-2C receptor, Serotonin receptor 2C |
| Gene Symbols | HTR2C |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.