Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 5-HT6 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | 5-HT6 |
---|---|
Clone | 2157A |
Dilution | Immunohistochemistry 1-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9496SP
![]() |
R&D Systems
MAB9496SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9496100
![]() |
R&D Systems
MAB9496100 |
100 μg |
Each for $467.50
|
|
|||||
Description
5-HT6 Monoclonal specifically detects 5-HT6 in Human samples. It is validated for Immunohistochemistry.Specifications
5-HT6 | |
Immunohistochemistry 1-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
5-hydroxytryptamine (serotonin) receptor 6,5-HT65-HT6R, HTR6, Serotonin receptor 6,5-HT-6,5-hydroxytryptamine receptor 6 | |
Human 5-HT6 peptide Accession # P50406 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human 5-HT6 in direct ELISAs. |
2157A | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P50406 | |
3362 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title